Optifit Pipette Tips
Supplier: Sartorius
790010 790011 790012 791000 791001 791002 791020 791021 791200 791202 791201 780304 780305 790351 790352 780300 780310 780308 LH-780316 791210 791211 783210 783211 790350 790200 790201 790203 790014 790013 790353 791003 791203 791213 791212 10033-048 10033-052 10033-060 10033-062 10033-064 LH-780316 791210 783210 783211 790013 790353 791003 791213 791212 790200 790201 790202 790203 790014 790010 790011 790012 791000 791001 791002 791020 791021 791200 791202 791201 780305 790351 790352 790350 780300 780310 LH-B790204 LH-B790354 LH-B791004 LH-B791024 LH-B791204 LH-B790204 LH-B790354 LH-B791004 LH-B791024 LH-B791204 780304 LH-780314 LHEB790204 LHEB790354 LHEB791004 LHEB791204 LH-X780200 LH-X780201 LH-X781000 LH-X781001 LH-X791000 LH-X791001
15000-454CS 242.94 CAD
15000-454 15000-456 15000-458 15000-460 15000-462 15000-464 15000-466 15000-468 15000-470 15000-472 15000-474 47744-628 47746-180 47747-284 47747-286 53495-368 53495-706 80077-466 89082-222 89100-008 89100-010 89100-012 89134-802 89134-804 89167-672 47747-294 89100-000 89100-006 89215-250 89215-252 89215-256 89217-380 89167-667 89167-671 89167-673 89167-675 89167-677 89167-679 89234-492 10033-048 10033-052 10033-060 10033-062 10033-064 CA89082-222 CA89100-000 CA89100-004 CA89100-006 CA89100-008 CA89100-010 CA89134-802 CA89134-804 CA89167-667 CA89167-671 CA89167-673 CA89167-677 CA89167-679 CA89215-250 CA89215-252 CA89215-254 CA89215-256 CA89217-380 CA89234-492 CA15000-454 CA15000-456 CA15000-458 CA15000-460 CA15000-462 CA15000-464 CA15000-466 CA15000-468 CA15000-470 CA15000-472 CA15000-474 CA47746-180 CA47747-284 CA47747-286 CA47747-294 CA53495-368 CA53495-706 10835-952 10835-954 10835-956 10835-958 10835-960 CA10835-952 CA10835-954 CA10835-956 CA10835-958 CA10835-960 CA47744-629 CA76019-342 76300-118 76300-120 76300-122 76300-124 CA76416-016 CA76416-018 CA76416-020 CA76416-022 CA77745-758 CA77745-760
Optifit Pipette Tips
Pipette Tips
Sartorius optifit pipette tips are an excellent choice for various laboratories and pipetting tasks with their wide range of packaging and purity options. These tips are non-filtered and designed for best possible fitting and sealing on Sartorius pipettes to secure the highest possible accuracy, precision, and reproducibility.
  • Ergonomic and light tip attachment and ejection secured by tip compatibility with the Optiload feature of Sartorius Picus® NxT, Picus®, eLINE®, mLINE® and Proline® Plus pipettes
  • Environmentally friendly materials used in tips, trays,
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable