Sulfite Test Kit, SU-5, Hach
Supplier: Hach
148002
CA1480-02EA 339.08 CAD
CA1480-02
Sulfite Test Kit, SU-5, Hach
Titrators
Utilizing drop count titration/iodometric to determine sulfite, this kit contains 100 tests.
  • Utilizes drop count titration - a simple, economical titration method
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Ordering information: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.