Chloroform

Supplier: Hach

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

1445817
CA14458-17EA 763.99 CAD
CA14458-17
Chloroform
Chloroform

ACS Grade, CAS No. 67-66-3 CHCl3, FW = 119.38.Assay: ≥99.8% 4L.

Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: -63
MDL Number: MFCD00000826
CAS Number: 67-66-3
UN: 1888
ADR: 6.1 III
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR