Sulfuric acid 0.1600 N, cartridge for digital titrator

Supplier: Hach
Danger

1438801
CA14388-01EA 64.2 CAD
CA14388-01
Sulfuric acid 0.1600 N, cartridge for digital titrator
Sulfuric acid

For determination of alkalinity by Digital Titrator titration.


  • Accurate results when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical


Hach method 8203. 0.1600 N Sulfuric Acid. Digits = mg/L CaCO3 in 100 mL, Digits x 0.002 = meq. Approximately 40-100 tests.

Formula: H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 3264
ADR: 8,II

Order Now

Specification Test Results

EPA compliant Yes
Method 8203
Method Name Phenolphthalein
Number of tests 40 - 100
Parameter Alkalinity
Platform Digital Titrator
Range 10 - 160 mg/L CaCO3
Titrant 0.1600 N H2SO4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR