Uvex Astrospec 3000® Safety Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
S1359 S1169 S1359C S1169C S135
CA14227-708EA 16.41 CAD
CA14227-708 CA26902-847 CA33001-996 CA33002-033 CA33002-037 CA33002-049 CA33002-051 CA33002-210 CA33002-265 33002-033 33002-036
Uvex Astrospec 3000® Safety Eyewear, Honeywell Safety
Safety Glasses
Unique shape and style provide excellent eye protection in a variety of frames and lenses.
  • Wide viewing area provides good visibility and protection from flying particles
  • Nylon frame and lens with molded-in side shields and brow guard
  • Four-stop temple adjustment (130 to 150 mm) provides a face-hugging fit
  • Optically correct, one-piece polycarbonate lens is lightweight and impact-resistant
  • Lens angle adjusts for a comfortable fit
  • Temples will not scratch the lens when folded
Models with Duoflex™ temples have soft, behind-the-ear comfort cushions.More Product Information