Short-Term Detector Tubes, Draeger
Supplier: Draeger
8101941 8101231 8101891 8101021 8101681 8101511 CH20501 CH25101 8101811 CH25601 CH24101 CH22901 8101121 6728071 CH31401 CH29901 6728051 8101141 6728761 6728451 8101351 6728961 6722101 CH31901 CH23201 CH20701 6730501 6728111 CH20201 6728381 6722701 CH29501 6728181 CH30301 6726665 8101711 8101741 6733081 CH31801 CH25001 6728521 CH20301 6726801 6728241 6728681 6718701 6733171 CH20401 CH19801 8101751 8101041 6728861 8101491 CH23501 CH20601 CH24301 6733051 6718901 8101601 6733231 CH19701 8103381
CA13453-106PK 580.82 CAD
CA13453-106 CA13453-110 CA13453-120 CA13453-132 CA13453-140 CA13453-142 CA13453-202 CA13453-206 CA13453-208 CA13453-212 CA13453-214 CA13453-238 CA13453-240 CA13453-248 CA13453-250 CA13453-252 CA13453-256 CA13453-258 CA13453-288 CA13453-290 CA13453-292 CA13453-294 CA13453-296 CA13453-298 CA13453-302 CA13453-306 CA13453-308 CA13453-314 CA13453-316 CA13453-318 CA13453-320 CA13453-322 CA13453-326 CA13453-328 CA13453-330 CA13453-354 CA13453-356 CA13453-358 CA13453-360 CA13453-362 CA13453-372 CA13453-374 CA13453-376 CA13453-384 CA13453-386 CA13453-408 CA13453-410 CA13453-414 CA13453-416 CA13453-420 CA13453-424 CA13453-430 CA13453-442 CA13453-138 CA13453-204 CA13453-210 CA13453-216 CA13453-254 13453-310 13453-172 13453-246 13453-378 89027-028
Short-Term Detector Tubes, Draeger
Gas Detection Systems
For use with the accuro® Gas Detector Pump (see 34330-068 series) and Aerotest Breathing Air Tester (see 34330-078).
  • Rapid and accurate readings
  • Instantaneous measurement of hazardous substances
For rapid and accurate readings, these tubes serve the targeted measurement of instantaneous concentrations of hazardous substances and the short-term measurements are carried out manually with pump unit.
Ordering information: For additional tubes, contact your Avantor sales representative. Halogenated hydrocarbons detector tubes (13453-172) must be shipped via motor freight.
Order Now
SPECIFICATIONS
Environmentally Preferable