DuPont™ Tyvek® 400 Coveralls with Elastic Wrists and Ankles, Zipper Front Closure, Comfort Fit Design
Supplier: DuPont
TY125SWHLG002500 TY125SWHXL002500 TY125SWH3X002500 TY125SWH5X002500 TY125SWH6X002500 TY125SWH7X002500 TY125SWHMD002500 TY125SWH2X002500
CA13452-874CS 1381.8 CAD
CA13452-874 CA13453-572 CA13453-576 CA47731-180 CA47731-182 CA47731-184 CA13452-872 CA13453-574
DuPont™ Tyvek® 400 Coveralls with Elastic Wrists and Ankles, Zipper Front Closure, Comfort Fit Design
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Comfort fit design
  • Laydown collar
  • Zipper front
  • Elastic wrists and ankles
  • Elastic waist
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 40More Product Information