Dupont™ ProShield® 60 Coveralls with Standard Hood and Attached Skid Resistant Boots
Supplier: DuPont
NG122SWHSM002500 NG122SWHMD002500 NG122SWHLG002500 NG122SWHXL002500 NG122SWH2X002500 NG122SWH3X002500 NG122SWH4X002500 NG122SWH5X002500 NG122SWH6X002500 NG122SWHSM002500 NG122SWH6X002500
CA13452-282CS 1264.05 CAD
CA13452-282 CA13452-284 CA13452-286 CA13452-288 CA13452-290 CA13452-292 CA13452-294 CA13452-296 10016-104 10016-106 CA10016-104
Dupont™ ProShield® 60 Coveralls with Standard Hood and Attached Skid Resistant Boots
Disposable Garments
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Standard fit hood
  • Elastic wrists
  • Attached skid-resistant boots
  • Serged seams
ProShield® 60 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Attached hood with elastic around face opening is standard fit. Front zipper closure allows for easy donning and doffing. Elastic openingMore Product Information