Chip Measurement System (CMS) Analyzer, Draeger
Supplier: Draeger
6406340 6406190 6406210 6406230 6406320 6406180 6406460 6406400 6406200 6406430 6406490 6406240 6406380 6406360 6406390 6406220 6406440 6406370 6406310 6406290 6406470 6406330 6406580 6406080 6406050 6406070 6406060 6406020 6406130 6406010 6406150 6406120 6406110 6406100 6406040 6406090 6406140 6406160 6406280 6406260 6405300 6406560 6406550 6406520 6406510 6406030 6406250
CA12777-224EA 1034.99 CAD
CA12777-224 CA12777-226 CA12777-228 CA12777-230 CA12777-232 CA12777-234 CA12777-236 CA12777-238 CA12777-240 CA12777-242 CA12777-244 CA12777-246 CA12777-248 CA12777-250 CA12777-252 CA12777-254 CA12777-256 CA12777-258 CA12777-260 CA12777-262 CA12777-264 CA12777-266 CA12777-268 CA12777-270 CA12777-272 CA12777-274 CA13450-868 CA34330-008 CA34330-010 CA34330-012 CA34330-014 CA34330-016 CA34330-018 CA34330-020 CA34330-022 CA34330-024 CA34330-026 CA34330-028 CA34330-030 CA34330-032 CA34330-034 CA34330-038 CA34331-678 CA34331-680 CA34331-682 CA34331-684 CA47729-678 CA47729-680 CA47729-686 CA47729-688 CA80080-910 CA34330-036 CA34330-824
Chip Measurement System (CMS) Analyzer, Draeger
Gas Detection Systems
This easy-to-use, portable system integrates a variety of substance-specific chemical sensing chips with a handheld reader.
  • Portable, easy to use
  • Integrates variety of substance-specific, chemical-sensing chips with handheld reader
Flow sensor guarantees a constant mass flow of the sample through the capillary in the chip so that measurements are not affected by fluctuations in the atmospheric pressure. It takes gas and vapor spot measurements by combining chemistry and electronics to provide accurate and reliable results.
Ordering information: More Product Information