Anti-CD229 (Ly-9) Armenian Hamster Monoclonal Antibody (Biotin) [clone: Ly9ab3]

Supplier: Biolegend
122903
CA122903-BLEA 132.45 CAD
CA122903-BL
Anti-CD229 (Ly-9) Armenian Hamster Monoclonal Antibody (Biotin) [clone: Ly9ab3]
Antibodies
Biotin anti-mouse CD229 (Ly-9) [Ly9ab3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD229 (Ly-9)
Clonality: Monoclonal
Clone: Ly9ab3
Conjugation: Biotin
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR