Electrical Lockout Pouch, Brady Worldwide®
Supplier: Avantor
CA12222-708EA 153.35 CAD
CA12222-708
Electrical Lockout Pouch, Brady Worldwide®
Bags Pouches and Belt Cases
Lightweight, portable pouch contains lockout devices to secure electrical equipment.
  • Metal loops allow for hanging on a belt
Pouch contents include; one 2" blue padlock, one red labeled lockout hasp, one universal multi pole breaker lockout, one 277 Volt 'No Hole' circuit breaker lockout, one small red plug lockout, and two heavy-duty lockout tags.