Lockout Tags, Brady Worldwide®
Supplier: Brady Worldwide
65526 65527 66060 66065 66061 66062 65503 65504 65505 65535 66063 66064 66054 66055 66066 66067 65506 65521 65522 65523 65670 66053 65534 65515 65524 65525 50198 50199 50250
CA12222-006PK 84.69 CAD
CA12222-006 CA12222-008 CA12222-012 CA12222-014 CA12222-016 CA12222-018 CA12222-022 CA12222-024 CA12222-026 CA12222-028 CA12222-042 CA12222-044 CA12222-058 CA12222-060 CA12222-062 CA12222-074 CA12222-076 CA12222-078 CA12222-104 CA12222-106 CA12222-108 CA12222-110 CA12222-116 CA12222-118 CA12222-148 CA12222-150 CA12222-152 CA12222-156 CA89007-152 CA89007-154 CA89007-158
Lockout Tags, Brady Worldwide®
Security Locks
Constructed of durable polyester, these lockout tags exceed OSHA's 50lb
  • Rigid polyester surface is easy to write on and stays legible longer than cardstock
  • 3/8" diameter brass eyelet accepts most padlock shackles
  • Tag size is 5.75"Hx3"W
  • Standard package includes 25 tags and nylon fasteners
pull requirement for tagout devices.