SYPRO® Ruby Protein Blot Stain, Lonza
Supplier: LONZA WALKERSVILLE INC. CA
50565
CA12001-746EA 446.16 CAD
CA12001-746
SYPRO® Ruby Protein Blot Stain, Lonza
Stains Electrophoresis Stains
SYPRO™ Ruby is 60-times more sensitive than reversible protein blot stains like Ponceau S, and 30-times more sensitive than Amido Black or Coomassie® brilliant blue stains.
  • Visualize proteins prior to transfer - does not interfere with protein activity
  • Versatile - Excellent for staining native proteins
  • Safe - No acids or organic solvents necessary
  • Sensitive - Detects as little as 4 to 8 ng protein per band