Sodium dodecyl sulfate (SDS) 10% in aqueous solution, AccuGENE®

Supplier: LONZA WALKERSVILLE INC. CA

51213
CA12001-390EA 110.51 CAD
CA12001-390 CA12001-392
Sodium dodecyl sulfate (SDS) 10% in aqueous solution, AccuGENE®
Sodium dodecyl sulphate (SDS)

AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.


  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs


Each reagent is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.


Sodium dodecyl sulfate or sodium lauryl sulfate (SDS/SLS, respectively) is an anionic surfactant commonly used in polyacrylamide gel electrophoresis for the determination of protein molecular weight. In most proteins, the binding of SDS to the polypeptide chain imparts an even distribution of charge per unit mass, thereby resulting in a fractionation by approximate size during electrophoresis. SDS solutions may also be used in nucleic acid extraction procedures for the disruption of cell walls and dissociation of nucleic acid:protein complexes

Formula: CH₃(CH₂)₁₁OSO₃Na
Density: 1…1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a941b1d45a998148141ae9d836c27824416a59f&vid=c681a5fe2d2a21eb8dd3c1a4deaaa933668ffb5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR