Clear Weight Handling Forceps, Mettler Toledo
Supplier: Mettler Toledo
30213633
CA11238-246EA 90.4 CAD
CA11238-246
Clear Weight Handling Forceps, Mettler Toledo
Forceps
Used for weight handling between 1 mg to 200 g.