TECHREF® H-2358 Sample Cells Single-Open Ended
Supplier: Tech Ref
H2358
CA11100-008PK 117.65 CAD
CA11100-008
TECHREF® H-2358 Sample Cells Single-Open Ended
Sample Containers Sample Cups for Analyzers
TECHREF® H-2358 Sample Cells can be used in all models of Horiba and single-load XOS XRF analyzers.
  • Designed with rings that allow the film/membrane to be seated within the sample cell
  • 100% inspected
  • Intended for one-time use and are fully disposable
  • There are two fill lines (5 and 10 ml)
TECHREF® H-2358 Sample Cells were originally designed specifically for use with the Carousel/Turntable models of Horiba XRF Analyzers to reduce the amount of dust and/or debris that can be picked up when rotating in the Carousel/Turntable unit, leading to moreMore Product Information