Combitips® advanced Positive Displacement Pipette Tips, PCR Clean
Supplier: Eppendorf
0030089766 0030089774 0030089782 0030089790 0030089804 0030089812 0030089820 0030089839 0030089847 0030089766 0030089774 0030089782 0030089790 0030089804 0030089812 0030089820 0030089839 0030089847
89510-208CS 446.05 CAD
89510-208 89510-210 89510-212 89510-214 89510-216 89510-218 89510-220 89510-222 89511-338 CA11033-306 CA11033-308 CA11033-310 CA11033-312 CA11033-314 CA11033-316 CA11033-318 CA11033-320 CA11033-322
Combitips® advanced Positive Displacement Pipette Tips, PCR Clean
Pipette Tips
Eppendorf Combitips® advanced in 'PCR clean' grade offer superior positive displacement pipetting, ensuring precise handling of sensitive liquids in PCR and molecular biology, with contamination-free, ergonomic design for reliable lab results.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • 9 volume sizes (0.1 to 50 ml) offer a maximum range of dispensing volumes with increments as low as 100 nl
  • Certified to be free from human DNA, DNase, RNase, and PCR inhibitors that could
More Product Information