Optimum Nitrile Pink Glove Powder-Free, Medicom
Supplier: Avantor
CA11033-070CS 588 CAD
CA11033-070 CA11033-072 CA11033-074
Optimum Nitrile Pink Glove Powder-Free, Medicom
Gloves
Optimum Nitrile Gloves use a proprietary nitrile formulation to create a lighter glove with great tactility and enhanced barrier protection.
  • Not made with Natural Rubber Latex
  • Accelerator-Free
  • Ambidextrous
  • Beaded Cuff
  • Textured Fingertips
  • Non-Sterile
  • Single Use
By packing 250 gloves per box, Optimum gloves use less packaging and create less waste.
Hedy Nitrile gloves offer an exclusive 'Small-Medium' size for those hands that are a little larger than small and a little smaller than medium.
Ordering information: Packaged 250 gloves per dispenser gloves.