Aluminum acetate, dibasic stabilised, powder

Supplier: SPECTRUM CHEMICAL MFG CORP CA


Stabilization: Stabilised with boric acid

AL223-125GM AL223-500GM AL223-2.5KG
CA11032-090EA 361.99 CAD
CA11032-090 CA11032-092 CA11032-094
Aluminum acetate, dibasic stabilised, powder
Aluminum acetate, dibasic

Aluminum Acetate, Dibasic, Stabilized with Boric Acid, Powder is a salt that is used as a topical astringent to treat skin irritations from poison ivy, substances such as soaps and cosmetics and insect bites. Aluminum acetate is particularly helpful for foot problems including athlete’s foot, swelling, and excessive sweating. It is also used to treat ear canal infections. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available


GHS hazard : Health hazard

Formula: (CH₃CO₂)Al(OH)₂
Storage Temperature: Ambient
MDL Number: MFCD00010672
CAS Number: 7360-44-3

Order Now

Specification Test Results

Assay (Al₂O₃) Approx.30 %
Solubility (5%, Water) Complete With
Chloride (Cl) max. 0.01 %
Heavy Metals (As Pb) max. 0.005 %
Iron (Fe) max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR