Charts and Pens for 8" Dickson® Chart Recorders, Graphic Controls
Supplier: Graphic Controls
31437935 31270492 31169363 32020491 31437885 31328621 00808956 00166256 31439105 00479279 00166207 00166132 31301917 20740017 31085874 00540252 31089355 00166249 31438206 31088167 30604030 31438008 00553669 00910448 31438180 00166157 31438123 31089363 31437935 31270492 31169363 32020491 31437885 31328621 00808956 00166256 31439105 00479279 00166207 00166132 31301917 20740017 31085874 00540252 31089355 00166249 31438206 31088167 30604030 31438008 00553669 00910448 31438180 00166157 31438123 31089363
CA11032-020PK 127.5 CAD
CA11032-020 CA11032-022 CA11032-024 CA11032-026 CA11032-028 CA11032-030 CA11032-032 CA11032-034 CA11032-036 CA11032-038 CA11032-040 CA11032-042 CA11032-044 CA11032-046 CA11032-048 CA11032-050 CA11032-052 CA11032-054 CA11032-056 CA11032-058 CA11032-060 CA11032-062 CA11032-064 CA11032-066 CA11032-068 CA11032-070 CA11032-072 CA11032-074 11032-020 11032-022 11032-024 11032-026 11032-028 11032-030 11032-032 11032-034 11032-036 11032-038 11032-040 11032-042 11032-044 11032-046 11032-048 11032-050 11032-052 11032-054 11032-056 11032-058 11032-060 11032-062 11032-064 11032-066 11032-068 11032-070 11032-072 11032-074
Charts and Pens for 8" Dickson® Chart Recorders, Graphic Controls
Chart Recorders Charts for Recorders
Graphic Controls supplies a full line of charts and markers to fit all available Dickson® recorders
Graphic Controls charts and markers provide precise fit and function for all available Dickson® recorders. Our products deliver OEM performance at factory direct savings.
Versatile and durable, Dickson® recorders are used in a wide variety of applications and conditions measuring temperature, humidity, and pressure. Graphic Controls charts and markers perfectly complement these diverse applications and conditions to ensure your critical data is accurately recorded and preserved.
Order Now
SPECIFICATIONS
Environmentally Preferable