Anti-ACP1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-4640S
CA11029-054EA 229.01 CAD
CA11029-054
Anti-ACP1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: ACP1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Potato
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR