Equilibration Tubes, Cytiva
Supplier: Cytiva
80646779
CA11028-370PK 195.01 CAD
CA11028-370
Equilibration Tubes, Cytiva
Tubes Dialysis Tubes
For equilibrating IPG strips prior to second dimension.
Ordering information: Includes recommendation regarding equilibration solution volumes.