IntelliCAL™ Ammonia ISE Electrode, Hach
Supplier: Hach
ISENH318103
CA11028-138EA 2927.5 CAD
CA11028-138 89501-112
IntelliCAL™ Ammonia ISE Electrode, Hach
Electrodes ISE Electrodes
IntelliCAL™ ISENH3181 is a digital, combination gas-sensing ammonia ion selective electrode (ISE) with a replaceable membrane module, refillable outer body, double junction reference and built-in temperature sensor
  • Provides fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • User alerted when re-calibration is needed
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
Special features include easy-to-replace membrane modules.
The IntelliCAL ISENH3181 probe is available with a 1More Product Information