Sodium hydroxide standard solution 50%

Supplier: Hach
Danger

218049
CA11027-990EA 199.13 CAD
CA11027-990
Sodium hydroxide standard solution 50%
Sodium hydroxide

Multi-purpose sodium hydroxide solution.

Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR