SpecTainer™ Urine Containers, Polypropylene, Simport Scientific
Supplier: Simport Scientific
C566-60DOECO C566-90DOECO C566-120DOECO C566-60AQSECO C566-90AQSECO C566-120AQSECO C567-60DOECO C567-90DOECO C567-120DOECO C567-60AQSECO C567-120AQSECO C566-60Y C566-90Y C566-120Y C566-60CYS C566-90CYS C566-120CYS C567-60Y C567-90Y C567-120Y C567-90CYS C567-120CYS C566-90AQSECO C566-120AQSECO C567-120DOECO C567-60AQSECO C567-90AQSECO C567-120AQSECO C566-60Y C566-90Y C566-120Y C566-60CYS C566-90CYS C566-120CYS C567-60Y C567-90Y C567-120Y C567-60CYS C567-90CYS C567-120CYS C566-120DOECO C566-60DOECO C566-90DOECO
CA11027-808CS 427.92 CAD
CA11027-808 CA11027-810 CA11027-812 CA11027-814 CA11027-816 CA11027-818 CA11027-820 CA11027-822 CA11027-824 CA11027-826 CA11027-830 CA11027-832 CA11027-834 CA11027-836 CA11027-838 CA11027-840 CA11027-842 CA11027-844 CA11027-846 CA11027-848 CA11027-852 CA11027-854 89498-232 89498-234 89498-240 89498-242 89498-244 89498-246 89498-248 89498-250 89498-252 89498-254 89498-256 89498-258 89498-260 89498-262 89498-264 89498-266 89498-268 89498-270 76299-226 76299-228 76299-230
SpecTainer™ Urine Containers, Polypropylene, Simport Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Specifically designed for collection, transport, and storage of urine specimens, Simport Scientific's SpecTainer™ is a shatter-resistant polypropylene container that eliminates tthe problems of leakage and evaporation
The eco-friendly SpecTainer™ will biodegrade in less than 7 years when in constant contact with the soil. No heat, physical stress, oxygen, or sunlight is necessary. Containers do not contain any metals, nor leave any toxic residue. Available in two unique models, containers and caps are manufactured without the use of plasticisers or mold release agents. MaterialsMore Product Information