Phthalaldehyde, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

P1139-100G
CA11027-340EA 4589.99 CAD
CA11027-340
Phthalaldehyde, Reagent Grade
Phthalaldehyde

o-Phthalaldehyde, Reagent is a reagent in the analysis of amino acids and involved in the synthesis of heterocyclic compounds. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₈H₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00003335
CAS Number: 643-79-8
UN: 2923
ADR: 86.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR