Ethyl-(S)-(-)-lactate ≥98.0%

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate

E1187-56 E1187-55002
CA11024-324EA 4062.01 CAD
CA11024-324 CA11024-326
Ethyl-(S)-(-)-lactate ≥98.0%
Ethyl (S)-(-)-lactate

Ethyl L-Lactate, also known as lactic acid ethyl ester, is industrially produced from petrochemical stocks. It is a monobasic ester formed from ethanol and lactic acid and is used as a solvent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 154
Melting Pt: -25
Density: 1.031
Flash Pt: 53
Storage Temperature: Ambient
MDL Number: MFCD00004518
CAS Number: 687-47-8
UN: 1192
ADR: 3 III
Merck Index: 12,03850

Order Now

Specification Test Results

Assay 98.0 % min.
Color (APHA) max.20
Moisture max. 0.30 %
Acid Number max. 0.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR