Water, deionized VWR®

Supplier: VWR

96887-540 96887-700 96887-940
CA11024-174EA 238.8 CAD
CA11024-174 CA11024-176 CA11024-180 10806-022
Water, deionized VWR®
Water

This general-purpose water is purified by deionization. Water has a pH of 5.0 to 7.0. It passes the test for reducing potassium permanganate.


  • Conductivity: 2.5 µmhos max.

Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Specific conductance at 25°C 2.5 µmhos max.
pH at 25°C 5.0 - 7.0
Heavy metals (as Pb) 0.01 ppm max.
Silicates (as SiO2) 0.01 ppm max.
Substances reducing KMnO4 Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR