MAS-3 Furnaces, Anachemia
Supplier: A.R. Olympique
029-891-02 029-891-04 029-891-01 029-891-05 029-891-06 029-891-07 029-891-08 029-891-09
CA11023-364EA 128281.77 CAD
CA11023-364 CA11023-366 CA11023-368 CA11023-370 CA11023-372 CA11023-374 CA11023-376 CA11023-378
MAS-3 Furnaces, Anachemia
Ovens Furnaces (Ashing and Muffle Ovens)
The Mine Assay MAS-3 fire assay furnace, formerly known as “Williams & Wilson,” is well known in the mining industry all across North America, as well as in other locations worldwide.
  • High-quality firebrick and insulating refractories
  • Four non-metallic silicon carbide heating elements
  • Power and control accessories wired in base of furnace
  • Two digital temperature controllers
  • Manually operated door vent and chamber roof vent ensure adequate airflow through the chamber
Solid construction, durable features, and dependable, continuous operation haveMore Product Information