Calibration Standard, ICP Trace Metals, ERA
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
524
CA11022-716EA 1218.31 CAD
CA11022-716
Calibration Standard, ICP Trace Metals, ERA
Standards Environmental Analysis Standards
This all metals calibration standard can be used as initial as well as continuing calibration verification.
One 500 mL whole-volume bottle of this ICP trace metals calibration standard, preserved with HNO3 and HCl is ready to use.
Ordering information: This standard comes with a certificate of traceability and uncertainty. Calibration standards preserved with HNO3 and HCl are required to ship as hazardous materials.