Water Pollution Standard, QC Plus – Demand, ERA
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
4013
CA11022-666EA 370.95 CAD
CA11022-666
Water Pollution Standard, QC Plus – Demand, ERA
Standards Environmental Analysis Standards
ERA's QC Plus program includes environmental analytes at concentrations that reflect realistic levels of pollutants in industrial settings
One 24 mL screw-cap vial yields up to 1 L of this water pollution, QC Plus Demand standard after dilution.
Each sample level is designed for wastewater and industrial analysis. These QC samples are an asset to any quality assurance program because they enable you to test your internal systems to ensure that your equipment, methods and analysts are producing quality data.