L(+)-Cysteine ≥98%

Supplier: Spectrum Chemical Mfg. Corp.
Warning

CY110-06
CA11020-398EA 394 CAD
CA11020-398
L(+)-Cysteine ≥98%
L-Cysteine

L-Cysteine is biosynthesized in humans and a semi-essential amino acid. It is used most often used as a food additive.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: HSCH₂CH(NH₂)CO₂H
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.523 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4
EINECS: 200-158-2
Merck Index: 13,02810

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR