Calcium Standard Solution, 100 mg/L as Ca (NIST), Hach
Supplier: Hach
4457649
CA11016-996EA 127.27 CAD
CA11016-996
Calcium Standard Solution, 100 mg/L as Ca (NIST), Hach
Standards Cationic Standards Calcium Standards
For accuracy checks and meter calibrations in calcium determinations.