EGTA solution

Supplier: Hach
Danger

2229726
CA11016-884EA 65.76 CAD
CA11016-884
EGTA solution
EGTA (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)

For calcium and magnesium hardness determination by the Calmagite colorimetric method


Hach method 8030.


Number of tests: 100

Formula: C₁₄H₂₄N₂O₁₀
MDL Number: MFCD00004291
CAS Number: 67-42-5
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR