m-Endo Agar Plates, Hach
Supplier: Hach
2811615
CA11016-724PK 382.68 CAD
CA11016-724
m-Endo Agar Plates, Hach
Media for Microbiology Prepared Media for Microbiology
m-Endo broth prepared agar plates detect total coliform bacteria (colonies with a greenish-gold sheen)
For potable water samples, confirm with Lauryl Tryptose and Brilliant Green Bile Broth tubes. Samples that test positive for total coliform bacteria should be further analyzed for the presence of fecal coliform or E. coli using EC Medium or EC Medium with MUG respectively.