Sodium thiosulfate 0.02256 N stabilised, cartridge for digital titrator

Supplier: Hach

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

2409101
CA11016-466EA 60.29 CAD
CA11016-466
Sodium thiosulfate 0.02256 N stabilised, cartridge for digital titrator
Sodium thiosulphate

For determination of Chlorine by Digital Titrator titration.


  • Accurate results - when and where you need them
  • Fast and Flexible
  • Versatile
  • Economica


0.02256 N Digits x 0.01 = mg/L CI2 in 100 mL, Digits x 0.0000282 = meq. Stabilized sodium thiosulfate formulatio

Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
Merck Index: 12,08844

Order Now

Specification Test Results

Method 8209
Method Name Iodometric
Number of tests 40 - 100
Parameter Chlorine, Total
Platform Digital Titrator
Range 1 - 400 mg/L Cl2
Titrant 0.02256 N Na2S2O3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR