Stablcal® Calibration Standard, 100 mNTU, Hach
Supplier: Hach
2723353
CA11016-444EA 594.85 CAD
CA11016-444
Stablcal® Calibration Standard, 100 mNTU, Hach
Standards Turbidity Standards
Calibration Standard, 2 point calibration. Stablcal®, 100 m NTU.
  • Precise, pre-mixed formazin solutions
  • Stablcal is Safe to Handle
  • No special glassware or difficult lab techniques
  • Developed for use in any turbidimeter.
  • Enhanced stability saves time and labor