Carbon Dioxide Test Kit, Model CA-DT, Hach
Supplier: Hach
2064100
CA11014-682EA 786.9 CAD
CA11014-682
Carbon Dioxide Test Kit, Model CA-DT, Hach
Titrators
Utilizing the Hach Digital Titrator, the Model CA-DT kit is used to determine carbon dioxide concentrations.
  • Hach Digital Titrator offers higher levels of precision and accuracy than drop count procedures, with reproducibility comparable to a buret titration
  • Rugged titration device is ideal for field use, with no glass burets required
Kit includes Digital Titrator, reagents and labware, manual and carrying case. Range: 10.1-1,000 mg/L. Contains approximately 50 tests.
Ordering information: Hazardous Items: Items with this mark may be consideredMore Product Information
Order Now
SPECIFICATIONS
Measuring range 10.1 - 1,000 mg/L CO2
Environmentally Preferable
Method Digital Titrator/Sodium hydroxide phenophthalein
Parameters Carbon Dioxide - As CO2
Weight 3.5 lbs (shipping weight)