Manganese Test Kit, Model MN-5, Hach
Supplier: Hach
146700
CA11014-452EA 467.44 CAD
CA11014-452
Manganese Test Kit, Model MN-5, Hach
Environmental Test Kits
Manganese causes a bitter taste in water, and at concentrations above 0.1 mg/L, it causes objectionable stains on laundry and plumbing fixtures.
  • Simple, lowcost color disc method is the most accurate visual test kit method
  • Achieve fast results with an easytocarry test kit
Manganese control also is important in the beverage, paper, and textile industries, and in dye production and food processing. An economical solution, this kit uses the simple, accurate color disc method.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.