Accessories for Digital block reactor, DRB 200, Hach
Supplier: Hach
DRB200-03 DRB200-01 DRB200-02 HHA155
CADRB200-03EA 4559.37 CAD
CADRB200-03 CA11014-306 CA11014-312 CA11024-998 89501-094 75919-344 75919-346 75919-348 75919-350
Accessories for Digital block reactor, DRB 200, Hach
Digestion Systems Mineralisation Workstations
This adapter converts the 20 mm diameter vial wells to 16 mm diameter.