Whatman™ Puradisc Syringe Filters, Glass Microfiber, Whatman products (Cytiva)
Supplier: Cytiva
6806-1316 6820-1316 6822-1312 6823-1327 6825-1307 6783-2510 6783-2520 6825-2527 6792-2510 6787-2520 6818-1304 6821-1310 6825-2517 6825-2517 6806-1316 6820-1316 6821-1310 6822-1312 6823-1327 6825-1307 6783-2510 6783-2520 6825-2527 6792-2510 6787-2520 6818-1304
15001-360PK 2260.51 CAD
15001-360 28137-566 28137-570 28137-572 28137-574 28137-576 28137-950 28137-952 28137-969 52857-999 70240-152 89022-362 28137-568 CA11013-914 10035-512 CA15001-360 CA28137-566 CA28137-568 CA28137-570 CA28137-572 CA28137-574 CA28137-576 CA28137-950 CA28137-952 CA28137-969 CA52857-999 CA70240-152 CA89022-362
Whatman™ Puradisc Syringe Filters, Glass Microfiber, Whatman products (Cytiva)
Filters Syringe Filters
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
  • Eight different material options
  • Choice of 4, 13, and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
  • Recyclable after use
Puradisc syringe filters cater to a wide variety of sample types andMore Product Information