Whatman™ Cellulose Nitrate Membranes
Supplier: Cytiva
7184-004 7184-014 7182-004 7184-009 7182-002 7184-002 7190-004 7193-004 7195-004 10400012 10401312 10400106 10400206 7182-009 7195-009 7193-002 7195-002 10402012 10400212 7181-004 7184-001 7188-004 10400112 10401112 7188-004 10400112 7182-009 10401312 10400106 7184-001 10401112 10400206 7193-002 7195-002 10402012 10400212 7195-009 10400012 10401126 10401118 10401106 10401170 10405079 7184-005 7182-001 10400114 10400014 10401314 10401121 7190-002 7191-014 7191-005 10400214 10401114 10402014 7186-004 7188-003 10400214 10401106 10401114 10401118 10401121 10401314 10402014 10405079 7182-001 7184-005 7186-004 7188-003 7190-002 7191-005 7191-014 10400014 10400114 10401126 7182-014 7188-009 7181-002 7188-002 7181-002 7182-014 7188-002 7188-009 7184-004 7184-014 7182-004 7184-009 7181-004 7182-002 7184-002 7190-004 7193-004 7195-004
28159-547PK 333.22 CAD
28159-547 28159-555 28159-570 28297-778 28454-042 28454-053 28454-079 28454-089 28454-097 CA28152-824 CA28152-825 CA28152-832 CA28152-834 CA28297-764 CA28297-802 CA28454-084 CA28454-093 CA74300-256 CA89014-304 28454-027 CA28297-774 28159-585 CA11008-540 CA11008-546 CA11008-588 11008-540 28297-764 28152-825 28152-832 28297-774 11008-546 28152-834 28454-084 28454-093 74300-256 89014-304 28297-802 28152-824 CA11012-716 CA11012-718 CA11012-726 CA11012-734 CA11013-034 CA11013-222 CA11013-224 CA11013-468 CA11013-470 CA11013-576 CA11013-582 CA11013-622 CA11013-628 CA11013-636 CA11013-884 CA11013-904 CA11013-960 CA11014-002 CA11014-034 10034-874 10034-896 10034-898 10034-900 10034-902 10034-904 10034-930 10034-956 10035-534 10035-536 10035-538 10035-542 10035-544 10035-546 10035-548 10035-690 10035-692 10035-694 10147-928 10147-932 10147-926 10147-930 CA10147-926 CA10147-928 CA10147-930 CA10147-932 CA28159-547 CA28159-555 CA28159-570 CA28297-778 CA28454-027 CA28454-042 CA28454-053 CA28454-079 CA28454-089 CA28454-097
Whatman™ Cellulose Nitrate Membranes
Filters Membrane Filters
Cellulose nitrate membranes, circles, plain. Recommended for the majority of routine applications, this membrane is manufactured under strictly controlled conditions.
  • Narrow pore size distribution for improved surface capture and analysis
  • Low levels of extractables to ensure sample integrity
The user will benefit from the performance improvements that are now available in Whatman membrane filters, including very narrow pore size distribution and low levels of extractables. Most membranes are inherently brittle and difficult to handle; it is notMore Product Information