Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.45 µm
Supplier: Cytiva
7141-154 7153-104 7141-104 7141-114 7141-124 10407332 10407312 10407372 10407370 10407334 10407324 10407314 10401670 7140-104 10406870 10407332 10407312 10407334 10401670 10406870 10407370 10407324 10407314 7140-104 10407372 10409772 10409470 10409472 10409772 10409470 10409472 10409771 10407970 10409770 10401672 10406572 10406872 10406871 10401672 10406572 10406871 10406872 10407970 10409771 10409770 7141-154 7153-104 7141-104 7141-114 7141-124
14230-030PK 1049.2 CAD
14230-030 28159-525 28297-732 28297-734 30253-266 CA13500-162 CA13500-170 CA28151-138 CA28151-140 CA28151-142 CA28151-146 CA28151-150 CA97016-792 CA11008-530 CA11008-580 13500-162 13500-170 28151-142 97016-792 11008-580 28151-140 28151-146 28151-150 11008-530 28151-138 89233-764 89233-766 89233-768 CA89233-764 CA89233-766 CA89233-768 CA11012-908 CA11013-500 CA11013-524 CA11013-584 CA11013-820 CA11013-980 CA11013-984 10034-918 10034-960 10034-966 10034-968 10034-970 10034-978 10035-800 CA14230-030 CA28159-525 CA28297-732 CA28297-734 CA30253-266
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.45 µm
Filters Membrane Filters
Whatman mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
  • Sterile options available for critical applications
  • Excellent contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have a mix of cellulose nitrate and cellulose acetate
  • The membrane offers a high degree of internal surface area for greater adsorption
More Product Information