Whatman™ Grade 91 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Cytiva
1091-165 1091-110 1091-125 1091-930 1091-190 1091-240 1091-185 1091-935 1091-150 1091-110 1091-125 1091-150 1091-165 1091-185 1091-190 1091-240 1091-930 1091-935
CA11013-286PK 477.12 CAD
CA11013-286 CA11013-288 CA11013-292 CA11013-304 CA11013-308 CA11013-310 CA11013-314 CA11013-316 CA11013-364 10035-238 10035-240 10035-700 10035-702 10035-704 10035-706 10035-708 10035-710 10035-712
Whatman™ Grade 91 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Papers Qualitative Filter Paper
A general purpose creped cellulose filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration.
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc.