Whatman™ Grade 604½ Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Cytiva
10312751 10312745 10312747 10312753 10312744 10312744 10312745 10312747 10312751 10312753
CA11012-766PK 185.64 CAD
CA11012-766 CA11012-768 CA11012-774 CA11012-780 CA11012-782 10034-712 10034-714 10034-716 10034-718 10034-720
Whatman™ Grade 604½ Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Papers Qualitative Filter Paper
Grade 604½ Qualitative Filter Papers for coarse precipitates. Maintains flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.
  • Increased total loading capacity as more filter area is available
Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.
Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.