Whatman™ Application-Specific Filter Papers, Grade 3000, Whatman products (Cytiva)
Supplier: Cytiva
10316114 10316114
CA11012-686PK 403.36 CAD
CA11012-686 10034-740
Whatman™ Application-Specific Filter Papers, Grade 3000, Whatman products (Cytiva)
Filters Filter Papers Qualitative Filter Paper
Smooth filter papers with good retentivity at a relatively high filtration speed. They are used for the clarifying filtration of dried beet pulp extracts or beet juice after the addition of lead acetate for subsequent polarimetric sugar determination.