SmartCal™ Moisture Analyzer Test Substance
Supplier: Mettler Toledo
30005791 30005793 30005790 30005792 30005918 30005917
CA11010-728EA 2333.3 CAD
CA11010-728 CA11010-730 CA11010-732 CA11010-734 CA11010-736 CA11010-738
SmartCal™ Moisture Analyzer Test Substance
Services
SmartCal™ is an innovative test substance that provides a fast way to verify the performance of a moisture analyzer.
  • Simply pour SmartCal™ into the sample pan and start
  • Test takes just 10 minutes
  • Easy to monitor and interpret instrument's performance over time
  • Store results directly in the instrument or manually enter into a free validated Excel® report sheet
  • Done just like a normal moisture measurement
  • Certified version available to satisfy regulatory requirements
  • SmartCal™ to get you up and running right away
SmartCal™ is a granular test substance.More Product Information