Omni Tip™ Homogenizing Kits, Omni International, Inc.
Supplier: Omni International
TH115-PCR TH115-PCRD TH115-PCR5D TH115-PCR7D TH115-PCR10D TH115-PCRH TH115-PCR5H TH115-PCR7H TH115-PCR10H
CA11010-514EA 8134.91 CAD
CA11010-514 CA11010-516 CA11010-518 CA11010-520 CA11010-522 CA11010-524 CA11010-526 CA11010-528 CA11010-530
Omni Tip™ Homogenizing Kits, Omni International, Inc.
Homogenizers Handheld Homogenizers
Omni Tip™ Homogenizing Kits include everything you need to homogenize
Patented Omni Tip™ Plastic Homogenizer Probes are designed to completely avoid cross-contamination between samples during homogenization. Engineered from a durable plastic, they come in two different configurations for soft tissue and hard tissue homogenizing. Omni Tips are available in a standard size of 7mm (Diameter) x 110mm (Length). The outer tube of Omni Tips is made of Lexan 144R, a polycarbonate. The inner shafts are made of Ultem 1000, a polyetherimide.
They come complete with an Omni TH Tissue HomogenizerMore Product Information