Heidolph® Titramax Vibrating Platform Shaker Package, Heidolph North America
Supplier: Heidolph NA, LLC
036130500CA
CA11009-588EA 16324.54 CAD
CA11009-588
Heidolph® Titramax Vibrating Platform Shaker Package, Heidolph North America
Shakers Microplate Shakers
The Vibrating Platform shaker is best used for research which requires ridged or gentle shaking for small samples on microtiter plates or test tubes
The Heating modules has a heating capacity of 300W which allows for quick temperature adjustments. Digital temperature setting up to 65C and separate display for actual temperature.
The Flat hood is for small vessels with a total height of 163mm, This is recommended especially for microtiter plates, culture plates, small flaks, media bottles and Erlenmeyer flasks up to 100ml. Transparent and non-fogging PETG material allows for sampleMore Product Information