Bench Top Sterilizers, 230 V, Heidolph Tuttnauer
Supplier: Heidolph NA, LLC
023210495CA 023210550CA
CA11009-238EA 34964.31 CAD
CA11009-238 CA11009-240 CA11009-254
Bench Top Sterilizers, 230 V, Heidolph Tuttnauer
Sterilizers Autoclaves (Steam Sterilizers)
Sterilizers provide a fast, safe, dependable, and convenient means of sterilizing wrapped, unwrapped, glass and tools. Not suitable for liquids or media (LabLine is suitable for liquids and media).
  • 230 V models
  • M models feature analog controls and gauges for simple operation
  • Standard features include a self-contained design that eliminates external plumbing and hardwiring for easy installation
  • Units include simple operation and fast and slow exhaust
  • ASME-stamped, 316 L, stainless steel chamber ensures reliability, durability, and high corrosion resistance
More Product Information